missing translation for 'onlineSavingsMsg'
Learn More

2-Butyrylthiophene, 97%, Thermo Scientific™

Product Code. 11437317
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437317 50 g 50g
11309579 5 g 5g
11427317 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437317 Supplier Thermo Scientific Alfa Aesar Supplier No. A15048.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5333-83-5
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00022503
InChI Key YXHIINNJOGKPLF-UHFFFAOYSA-N
Synonym 2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz
PubChem CID 79248
SMILES CCCC(=O)C1=CC=CS1

Specifications

Density 1.09
Boiling Point 242°C to 244°C
Flash Point 102°C (216°F)
Odor Characteristic
Refractive Index 1.543
Quantity 50 g
Beilstein 110167
IUPAC Name 1-thiophen-2-ylbutan-1-one
Formula Weight 154.23
Percent Purity 97%
Chemical Name or Material 2-Butyrylthiophene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.