accessibility menu, dialog, popup

UserName

H3COS

CAS RN 5333-83-5

IUPAC Name: 1-(thiophen-2-yl)butan-1-one
Synonyms: 1-(thiophen-2-yl)butan-1-one
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: YXHIINNJOGKPLF-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=CS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results