missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoquinoline, 98%

Product Code. 15595256 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15595256

Brand: Thermo Scientific Acros 457640050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 2005-43-8 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2005-43-8
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00234480
InChI Key QKJAZPHKNWSXDF-UHFFFAOYSA-N
Synonym quinoline, 2-bromo, bromoquinoline, pubchem21827, acmc-1cfcf, 2-bromoquinoline, ksc201o8j, 2-bromoquinoline 1g, abbypharma ap-12-10890
PubChem CID 2762756
IUPAC Name 2-bromoquinoline
SMILES BrC1=CC=C2C=CC=CC2=N1

Specifikationer

Melting Point 49.0°C to 51.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 2-Bromoquinoline
compliance-icons
Produktbeteckning
  • 2-Bromoquinoline
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Faroangivelser
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.