accessibility menu, dialog, popup

UserName

CAS RN 2005-43-8

BrN

CAS RN 2005-43-8

IUPAC Name: 2-bromoquinoline
Synonyms: 2-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: QKJAZPHKNWSXDF-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC=CC2=N1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)