Learn More
2-Bromophenyl isocyanate, 97%, Thermo Scientific Chemicals
£34.60 - £372.00
Chemical Identifiers
CAS | 1592-00-3 |
---|---|
Molecular Formula | C7H4BrNO |
Molecular Weight (g/mol) | 198.019 |
MDL Number | MFCD00001995 |
InChI Key | GOOVAYJIVMBWPP-UHFFFAOYSA-N |
Synonym | 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate |
PubChem CID | 137097 |
IUPAC Name | 1-bromo-2-isocyanatobenzene |
SMILES | C1=CC=C(C(=C1)N=C=O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11373206
|
Thermo Scientific Alfa Aesar
L12521.03 |
1 g |
£34.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383206
|
Thermo Scientific Alfa Aesar
L12521.06 |
5 g |
£94.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393206
|
Thermo Scientific Alfa Aesar
L12521.14 |
25 g |
£372.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1592-00-3 | |
198.019 | |
GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
137097 | |
C1=CC=C(C(=C1)N=C=O)Br |
C7H4BrNO | |
MFCD00001995 | |
2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
1-bromo-2-isocyanatobenzene |
Specifications
1592-00-3 | |
91°C to 92°C (5 mmHg) | |
Pungent | |
1.585 | |
UN2206 | |
Moisture sensitive | |
GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
1-bromo-2-isocyanatobenzene | |
137097 | |
97% |
1.607 | |
107°C (224°F) | |
C7H4BrNO | |
MFCD00001995 | |
3588447 | |
2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
C1=CC=C(C(=C1)N=C=O)Br | |
198.019 | |
198.03 | |
2-Bromophenyl isocyanate |
Safety and Handling
GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H331-H334-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only