missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl isocyanate, 97%

Product Code. 11393206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393206 25 g 25g
11373206 1 g 1g
11383206 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393206 Supplier Thermo Scientific Alfa Aesar Supplier No. L12521.14
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1592-00-3 | C7H4BrNO | 198.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-00-3
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD00001995
InChI Key GOOVAYJIVMBWPP-UHFFFAOYSA-N
Synonym 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate
PubChem CID 137097
IUPAC Name 1-bromo-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)Br

Specifications

Density 1.607
Boiling Point 91°C to 92°C (5 mmHg)
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.585
Quantity 25 g
UN Number UN2206
Beilstein 3588447
Sensitivity Moisture sensitive
Formula Weight 198.03
Percent Purity 97%
Chemical Name or Material 2-Bromophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.