missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-hydrazinopyridine, 98%

Product Code. 15481959
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481959 25 g 25g
15461959 1 g 1g
15471959 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481959 Supplier Thermo Scientific Alfa Aesar Supplier No. H64664.14
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 26944-71-8 | C5H6BrN3 | 188.028 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26944-71-8
Molecular Formula C5H6BrN3
Molecular Weight (g/mol) 188.028
MDL Number MFCD09743754
InChI Key PQMFVUNERGGBPG-UHFFFAOYSA-N
Synonym 2-bromo-6-hydrazinylpyridine, 2-bromo-6-hydrazinopyridine, 2-6-bromopyridin-2-yl hydrazine, 6-bromo-pyridin-2-yl-hydrazine, 2-bromo-6-hydrazinopydine, 6-bromo-2-pyridyl hydrazine, 6-bromopyridin-2-yl hydrazine, 6-bromo-2-hydrazino pyridine, 2 1h-pyridinone, 6-bromo-, hydrazone, pubchem12987
PubChem CID 19392588
IUPAC Name (6-bromopyridin-2-yl)hydrazine
SMILES C1=CC(=NC(=C1)Br)NN

Specifications

Melting Point 117°C to 118°C
Color Brown
Quantity 25 g
UN Number UN2811
Beilstein 386966
Formula Weight 188.03
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-hydrazinopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.