CAS RN 26944-71-8
IUPAC Name:
2-bromo-6-hydrazinylpyridine
Synonyms:
2-bromo-6-hydrazinylpyridine
Molecular Weight (g/mol):
188.03
Molecular Formula:
C5H6BrN3
InChi Key:
PQMFVUNERGGBPG-UHFFFAOYSA-N
SMILES:
NNC1=NC(Br)=CC=C1
1
–
1
of
1
results
1
2-Bromo-6-hydrazinopyridine, 98%
CAS: 26944-71-8 Molecular Formula: C5H6BrN3 Molecular Weight (g/mol): 188.028 MDL Number: MFCD09743754 InChI Key: PQMFVUNERGGBPG-UHFFFAOYSA-N Synonym: 2-bromo-6-hydrazinylpyridine,2-bromo-6-hydrazinopyridine,2-6-bromopyridin-2-yl hydrazine,6-bromo-pyridin-2-yl-hydrazine,2-bromo-6-hydrazinopydine,6-bromo-2-pyridyl hydrazine,6-bromopyridin-2-yl hydrazine,6-bromo-2-hydrazino pyridine,2 1h-pyridinone, 6-bromo-, hydrazone,pubchem12987 PubChem CID: 19392588 IUPAC Name: (6-bromopyridin-2-yl)hydrazine SMILES: C1=CC(=NC(=C1)Br)NN