missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chlorobenzaldehyde, 98%

Product Code. 15441889
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441889 250 mg 250mg
15421889 1 g 1g
15431889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441889 Supplier Thermo Scientific Alfa Aesar Supplier No. H64605.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64622-16-8 | C7H4BrClO | 219.462 g/mol

2-Bromo-6-chlorobenzaldehyde plays an important role as a linker, which provides higher selectivity and reactivity in the Buchwald C-N bond forming reaction in order to prepare Bruton's tyrosine kinase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-chlorobenzaldehyde plays an important role as a linker, which provides higher selectivity and reactivity in the Buchwald C-N bond forming reaction in order to prepare Bruton′s tyrosine kinase inhibitor.

Solubility
Slightly soluble in water. 

Notes
Store in a cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64622-16-8
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00266792
InChI Key NUGMENVSVAURGO-UHFFFAOYSA-N
Synonym 2-chloro-6-bromobenzaldehyde, benzaldehyde, 2-bromo-6-chloro, 2-bromo-6-chloro-benzaldehyde
PubChem CID 13524042
IUPAC Name 2-bromo-6-chlorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)C=O)Cl

Specifications

Melting Point 81°C to 86°C
Quantity 250 mg
Beilstein 2613087
Solubility Information Slightly soluble in water.
Formula Weight 219.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-chlorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.