accessibility menu, dialog, popup

UserName

BrOCl

CAS RN 64622-16-8

IUPAC Name: 2-bromo-6-chlorobenzaldehyde
Synonyms: 2-bromo-6-chlorobenzaldehyde
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: NUGMENVSVAURGO-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results