missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoroaniline, 99%, Thermo Scientific™

Product Code. 10400514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10400514 25 g 25g
10490504 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10400514 Supplier Thermo Scientific Acros Supplier No. 398840250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1003-99-2
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.01
MDL Number MFCD00070750
InChI Key FWTXFEKVHSFTDQ-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzenamine, 2-bromo-5-fluoro-aniline, 2-bromo-5-fluoro-phenylamine, 2-bromo-5-fluoro-benzenamine, 2-bromo-5-fluorophenylamine, pubchem1524, 5-fluoro-2-bromoaniline, 2-bromo-5-fluoro aniline, acmc-1c3a5
PubChem CID 2773317
IUPAC Name 2-bromo-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Br

Specifications

Melting Point 43°C to 47°C
Boiling Point 109°C to 110°C (15.0 mmHg)
Flash Point 110°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula BrC6H3(F)NH2
Quantity 25 g
Formula Weight 190.01
Percent Purity 99%
Chemical Name or Material 2-Bromo-5-fluoroaniline
compliance-icons
Product Identifier
  • 2-Bromo-5-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.