missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzenesulfonyl chloride, 97%

Product Code. p-7058515
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325555 1 g 1g
11335555 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325555 Supplier Thermo Scientific Alfa Aesar Supplier No. L19713.03
Add to basket
This item is not returnable. View return policy

CAS: 351003-45-7 | C6H3BrClFO2S | 273.5 g/mol

2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-45-7
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD03094149
InChI Key GJPWPFVLPNTOOL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzene-1-sulfonyl chloride, 2-bromo-4-fluorobenzenesulphonyl chloride, 2-bromo-4-fluorobenzenesulfonylchloride, 4-fluoro-2-bromobenzenesulfonyl chloride, 2-bromo-4-fluorophenyl chlorosulfone, benzenesulfonyl chloride, 2-bromo-4-fluoro, pubchem8367, acmc-209vnu, 2-bromo-4-fluoro-benzenesulfonyl chloride, ksc221s9l
PubChem CID 2778216
IUPAC Name 2-bromo-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)Br)S(=O)(=O)Cl

Specifications

Density 1.87
Boiling Point 264°C to 265°C
Refractive Index 1.578
Quantity 1 g
UN Number UN2920
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 273.51
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-fluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.