accessibility menu, dialog, popup

UserName

CAS RN 351003-45-7

IUPAC Name: 2-bromo-4-fluorobenzene-1-sulfonyl chloride
Synonyms: 2-bromo-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 273.5
Molecular Formula: C6H3BrClFO2S
InChi Key: GJPWPFVLPNTOOL-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results