missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxycinnamic acid, 97%, Thermo Scientific™

Product Code. 11462888
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462888 100 g 100g
11442888 5 g 5g
11452888 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462888 Supplier Thermo Scientific Alfa Aesar Supplier No. A16862.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151539-52-5
Molecular Formula C11H11BrO4
Molecular Weight (g/mol) 287.109
MDL Number MFCD00017614
InChI Key MSJCWIFRXCLULX-ONEGZZNKSA-N
Synonym 2-bromo-4,5-dimethoxycinnamic acid, 6-bromo-3,4-dimethoxycinnamic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, e-3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 2-propenoic acid, 3-2-bromo-4,5-dimethoxyphenyl, e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl-2-propenoic acid
PubChem CID 735828
IUPAC Name (E)-3-(2-bromo-4,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C(=C1)C=CC(=O)O)Br)OC

Specifications

Melting Point 253°C to 256°C
Quantity 100 g
Beilstein 3142451
Formula Weight 287.12
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,5-dimethoxycinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.