missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluoroanisole, 98%, Thermo Scientific™

Product Code. 11450447
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11450447

Brand: Thermo Scientific Alfa Aesar B21792.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceuticals.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-58-5
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.02
MDL Number MFCD00070753
InChI Key DKARISQFDBAJEZ-UHFFFAOYSA-N
Synonym 2-bromo-4,5-difluoroanisole, benzene, 1-bromo-4,5-difluoro-2-methoxy, 4-bromo-1,2-difluoro-5-methoxybenzene, pubchem5223, acmc-1co9o, 2-bromo4,5-difluoro-anisole, ksc495e7d, 2-bromo-4,5-difluoro-anisole, 1-bromo-4,5-difluoro-2-methoxy-benzene, 1-bromanyl-4,5-bis fluoranyl-2-methoxy-benzene
PubChem CID 2736262
IUPAC Name 1-bromo-4,5-difluoro-2-methoxybenzene
SMILES COC1=CC(F)=C(F)C=C1Br

Specifications

Quantity 2 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 223.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,5-difluoroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.