CAS RN 202865-58-5
CAS RN 202865-58-5
IUPAC Name:
1-bromo-4,5-difluoro-2-methoxybenzene
Synonyms:
1-bromo-4,5-difluoro-2-methoxybenzene
Molecular Weight (g/mol):
223.02
Molecular Formula:
C7H5BrF2O
InChi Key:
DKARISQFDBAJEZ-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=C(F)C=C1Br
1
–
1
of
1
results
1
2-Bromo-4,5-difluoroanisole, 98%, Thermo Scientific™
CAS: 202865-58-5 Molecular Formula: C7H5BrF2O Molecular Weight (g/mol): 223.02 MDL Number: MFCD00070753 InChI Key: DKARISQFDBAJEZ-UHFFFAOYSA-N Synonym: 2-bromo-4,5-difluoroanisole,benzene, 1-bromo-4,5-difluoro-2-methoxy,4-bromo-1,2-difluoro-5-methoxybenzene,pubchem5223,acmc-1co9o,2-bromo4,5-difluoro-anisole,ksc495e7d,2-bromo-4,5-difluoro-anisole,1-bromo-4,5-difluoro-2-methoxy-benzene,1-bromanyl-4,5-bis fluoranyl-2-methoxy-benzene PubChem CID: 2736262 IUPAC Name: 1-bromo-4,5-difluoro-2-methoxybenzene SMILES: COC1=CC(F)=C(F)C=C1Br