missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-nitropyridine, 98%, Thermo Scientific Chemicals

Product Code. 11334416
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19755-53-4
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00955613
InChI Key ZCCUFLITCDHRMG-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-pyridine, pyridine, 2-bromo-3-nitro, pyridine, 2-bromonitro, pubchem5955, acmc-1bt4y, 2-bromo-3-nitro pyridine, #, ksc490s2t, 2-bromo-3-nitropyridine
PubChem CID 555054
IUPAC Name 2-bromo-3-nitropyridine
SMILES [O-][N+](=O)C1=CC=CN=C1Br

Specifications

Melting Point 122°C to 125°C
Quantity 5 g
Beilstein 124504
Formula Weight 203
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'