accessibility menu, dialog, popup

UserName

ONONBr

CAS RN 19755-53-4

IUPAC Name: 2-bromo-3-nitropyridine
Synonyms: 2-bromo-3-nitropyridine
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: ZCCUFLITCDHRMG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CN=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results