missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10051264 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051264 5 g Glass bottle 5g
10205154 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051264 Supplier Thermo Scientific Acros Supplier No. 398860050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 53663-39-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
MDL Number MFCD00079721
InChI Key LSRTWJCYIWGKCQ-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid, 2-bromo-m-toluic acid, 2-bromo-3-carboxytoluene, 2-bromo-3-methylbenzoicacid, 3-methyl-2-bromobenzoic acid, benzoic acid, bromo-3-methyl, 2-brom-3-methylbenzoic acid, pubchem3805, acmc-1ay3t, ksc497o4r
PubChem CID 2735588
IUPAC Name 2-bromo-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1Br)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 135.0°C to 138.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H3(CH3)COOH
Quantity 5 g
Formula Weight 215.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromo-3-methylbenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.