accessibility menu, dialog, popup

UserName

CH3BrOHO

CAS RN 53663-39-1

IUPAC Name: 2-bromo-3-methylbenzoic acid
Synonyms: 2-bromo-3-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: LSRTWJCYIWGKCQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results