missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(2-tetrahydropyranyloxy)pyridine, 97%, Thermo Scientific™

Product Code. p-7059521
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460031 1 g 1g
11470031 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460031 Supplier Thermo Scientific Alfa Aesar Supplier No. 044774.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93560-60-2
Molecular Formula C10H12BrNO2
Molecular Weight (g/mol) 258.115
MDL Number MFCD06411700
InChI Key NXVQJPQYTUJVQP-UHFFFAOYSA-N
Synonym 2-bromo-3-oxan-2-yloxy pyridine, 2-bromo-3-tetrahydro-2-pyranyloxy pyridine, 2-bromo-3-tetrahydro-2h-pyran-2-yl oxy pyridine, 2-bromo-3-oxan-2-yl oxy pyridine, 2-bromo-3-2-tetrahydropyranyloxy pyridine, 3-2h-3,4,5,6-tetrahydropyran-2-yloxy-2-bromopyridine
PubChem CID 13319532
IUPAC Name 2-bromo-3-(oxan-2-yloxy)pyridine
SMILES C1CCOC(C1)OC2=C(N=CC=C2)Br

Specifications

Melting Point 67°C
Quantity 1 g
Formula Weight 258.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-3- (2-tetrahydropyranyloxy)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.