accessibility menu, dialog, popup

UserName

CAS RN 93560-60-2

OONBr

CAS RN 93560-60-2

IUPAC Name: 2-bromo-3-(oxan-2-yloxy)pyridine
Synonyms: 2-bromo-3-(oxan-2-yloxy)pyridine
Molecular Weight (g/mol): 258.12
Molecular Formula: C10H12BrNO2
InChi Key: NXVQJPQYTUJVQP-UHFFFAOYNA-N
SMILES: BrC1=C(OC2CCCCO2)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results