missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 10356744
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2011-06-5
CHEBI:28804
This item is not returnable. View return policy
2011-06-5
CHEBI:28804

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 44.0°C to 46.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H11NO2
MDL Number MFCD00008097
Quantity 1 g
Synonym d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape
InChI Key SNDPXSYFESPGGJ-SCSAIBSYSA-N
SMILES CCC[C@@H](N)C(O)=O
IUPAC Name (2R)-2-aminopentanoic acid
Molecular Weight (g/mol) 117.15
PubChem CID 439575
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material 2-Benzyloxy-3-methoxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzyloxy-3-methoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'