accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 2011-06-5

IUPAC Name: 2-(benzyloxy)-3-methoxybenzaldehyde
Synonyms: 2-(benzyloxy)-3-methoxybenzaldehyde
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: KXBGOVZWCRLLOR-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results