missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-1,3-dioxolane, 94%, Thermo Scientific™

Product Code. 10157243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157243 100 g Glass bottle 100g
10541144 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157243 Supplier Thermo Scientific Acros Supplier No. 388681000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 101-49-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00003215
InChI Key SSZACLYPEFCREM-UHFFFAOYSA-N
PubChem CID 7562
IUPAC Name 2-benzyl-1,3-dioxolane
SMILES C1COC(O1)CC2=CC=CC=C2

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0850g/mL
Boiling Point 115°C to 120°C (12.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.522
Quantity 100 g
Specific Gravity 1.085
Synonym 2-benzyldioxolan, 1,3-dioxolane, 2-phenylmethyl, phenylacetaldehyde glycol acetal, phenylacetaldehyde ethylene acetal, 1,3-dioxolane, 2-benzyl, phenylacetaldehyde ethyleneglycol acetal, phenylacetaldehyde ethylene glycol acetal, 1,3-dioxolane,2-phenylmethyl, benzyldioxolan, 1, 2-benzyl
Solubility Information Solubility in water: insoluble
Formula Weight 164.2
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2-Benzyl-1, 3-dioxolane
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzyl-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.