accessibility menu, dialog, popup

UserName

OO

CAS RN 101-49-5

IUPAC Name: 2-benzyl-1,3-dioxolane
Synonyms: phenylacetaldehyde ethyleneglycol cyclic acetal
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: SSZACLYPEFCREM-UHFFFAOYSA-N
SMILES: C(C1OCCO1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)