missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole, 97%

Product Code. 11419892
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419892 500 g 500g
11339679 50 g 50g
11409892 100 g 100g
11349679 250 g 250g
11429892 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11419892 Supplier Thermo Scientific Alfa Aesar Supplier No. A12026.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-50-4 | C3H4N2S | 100.14 g/mol

2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-50-4
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005325
InChI Key RAIPHJJURHTUIC-UHFFFAOYSA-N
Synonym 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
PubChem CID 2155
ChEBI CHEBI:40782
IUPAC Name 1,3-thiazol-2-amine
SMILES NC1=NC=CS1

Specifications

Melting Point 88°C to 92°C
Boiling Point 117°C (15 mmHg)
Quantity 500 g
Beilstein 105738
Merck Index 14,479
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 100.14
Percent Purity 97%
Chemical Name or Material 2-Aminothiazole
compliance-icons
Product Identifier
  • 2-Aminothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Thiazolamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.