Learn More
2-Aminothiazole, 97%
Description
2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
Specifications
Specifications
| Melting Point | 88°C to 92°C |
| Boiling Point | 117°C (15 mmHg) |
| Quantity | 250 g |
| Beilstein | 105738 |
| Merck Index | 14,479 |
| Solubility Information | Soluble in water,alcohol and ether. |
| Formula Weight | 100.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Aminothiazole |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.