missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-thien-2-ylbenzoic acid, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 209.4°C to 210°C |
| Color | Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Synonym | 2-amino-5-thiophen-2-yl benzoic acid, 2-amino-5-thien-2-ylbenzoic acid, 2-amino-5-2-thienyl benzoic acid, benzoicacid, 2-amino-5-2-thienyl, benzoicacid,2-amino-5-2-thienyl |
| Formula Weight | 219.26 |
| Show More |
Safety and Handling
Product Identifier
- 2-Amino-5-thien-2-ylbenzoic acid
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?