accessibility menu, dialog, popup

UserName

CAS RN 885268-32-6

SNH2OHO

CAS RN 885268-32-6

IUPAC Name: 2-amino-5-(thiophen-2-yl)benzoic acid
Synonyms: 2-amino-5-(thiophen-2-yl)benzoic acid
Molecular Weight (g/mol): 219.26
Molecular Formula: C11H9NO2S
InChi Key: PXSSPVWKCSZVKQ-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C=C1)C1=CC=CS1)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results