missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodopyridine-3-methanol, 97%, Thermo Scientific™

Product Code. 11464370
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11464370 5 g 5g
11454370 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11464370

Brand: Thermo Scientific Alfa Aesar H50011.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618107-90-7
Molecular Formula C6H7IN2O
Molecular Weight (g/mol) 250.04
MDL Number MFCD05663524
InChI Key PRUOIEDNVWNRSO-UHFFFAOYSA-N
Synonym 2-amino-5-iodopyridin-3-yl methanol, 2-amino-5-iodo-pyridin-3-yl-methanol, 2-amino-5-iodopyridine-3-methanol, 3-pyridinemethanol,2-amino-5-iodo, 2-amino-5-iodo-3-pyridinyl methanol, 2-amino-5-iodopyridin-3-yl-methanol, 2-amino-5-iodo-3-pyridyl methan-1-ol, pubchem7674, acmc-1b9kk, 3-pyridinemethanol, 2-amino-5-iodo
PubChem CID 17750179
IUPAC Name (2-amino-5-iodopyridin-3-yl)methanol
SMILES NC1=NC=C(I)C=C1CO

Specifications

Quantity 5 g
Sensitivity Light sensitive
Formula Weight 250.04
Percent Purity 97%
Chemical Name or Material 2-Amino-5-iodopyridine-3-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.