accessibility menu, dialog, popup

UserName

CAS RN 618107-90-7

NH2NIOH

CAS RN 618107-90-7

IUPAC Name: (2-amino-5-iodopyridin-3-yl)methanol
Synonyms: (2-amino-5-iodopyridin-3-yl)methanol
Molecular Weight (g/mol): 250.04
Molecular Formula: C6H7IN2O
InChi Key: PRUOIEDNVWNRSO-UHFFFAOYSA-N
SMILES: NC1=NC=C(I)C=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results