missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-diphenylthiazole, 97%, Thermo Scientific™

Product Code. 15498277
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498277 1 g 1g
15408287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498277 Supplier Thermo Scientific Alfa Aesar Supplier No. H51818.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne).

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6318-74-7
Molecular Formula C15H12N2S
Molecular Weight (g/mol) 252.335
MDL Number MFCD00438803
InChI Key XDWGEGZDMFHZBL-UHFFFAOYSA-N
PubChem CID 73222
IUPAC Name 4,5-diphenyl-1,3-thiazol-2-amine
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)N)C3=CC=CC=C3

Specifications

Odor Odorless
Quantity 1 g
Synonym 2-amino-4,5-diphenylthiazole, 2-thiazolamine, 4,5-diphenyl, 4,5-diphenyl-thiazol-2-ylamine, 4,5-diphenylthiazol-2-amine, diphenyl-1,3-thiazol-2-amine, 2-thiazolamine,4,5-diphenyl, thiazole, 2-amino-4,5-diphenyl, 4,5-diphenyl-2-thiazolamine, monohydrochloride, 4,5-diphenyl-1,3-thiazole-2-ylamine, ccris 8105
Solubility Information Insoluble in water. Soluble in chloroform and methanol.
Formula Weight 252.33
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5-diphenylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.