CAS RN 6318-74-7
IUPAC Name:
diphenyl-1,3-thiazol-2-amine
Synonyms:
diphenyl-1,3-thiazol-2-amine
Molecular Weight (g/mol):
252.34
Molecular Formula:
C15H12N2S
InChi Key:
XDWGEGZDMFHZBL-UHFFFAOYSA-N
SMILES:
NC1=NC(=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Amino-4,5-diphenylthiazole, 97%, Thermo Scientific™
CAS: 6318-74-7 Molecular Formula: C15H12N2S Molecular Weight (g/mol): 252.335 MDL Number: MFCD00438803 InChI Key: XDWGEGZDMFHZBL-UHFFFAOYSA-N PubChem CID: 73222 IUPAC Name: 4,5-diphenyl-1,3-thiazol-2-amine SMILES: C1=CC=C(C=C1)C2=C(SC(=N2)N)C3=CC=CC=C3