missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-3'-methoxyflavone, 99%

Product Code. 15466419
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466419 25 mg 25mg
15446419 5 mg 5mg
15456419 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466419 Supplier Thermo Scientific Alfa Aesar Supplier No. J64485.MB
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A MAP kinase inhibitor | CAS: 167869-21-8 | C16H13NO3 | 267.28 g/mol

A MAP kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167869-21-8
Molecular Formula C16H13NO3
Molecular Weight (g/mol) 267.28
MDL Number MFCD00671789
InChI Key QFWCYNPOPKQOKV-UHFFFAOYSA-N
Synonym 2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone
PubChem CID 4713
ChEBI CHEBI:77954
IUPAC Name 2-(2-amino-3-methoxyphenyl)chromen-4-one
SMILES COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 267.28
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Amino-3'-methoxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.