missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%

Product Code. 15435267
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15435267

Brand: Thermo Scientific Alfa Aesar H32532.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 852062-17-0 | C6H4F4N2 | 180.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 852062-17-0
Molecular Formula C6H4F4N2
Molecular Weight (g/mol) 180.106
MDL Number MFCD10699117
InChI Key HAVVQRHBCUBQGG-UHFFFAOYSA-N
Synonym 2-amino-3-fluoro-5-trifluoromethyl pyridine, 3-fluoro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-fluoro-5-trifluoromethylpyridine, 3-fluoro-5-trifluoromethyl-2-pyridylamine, 3-fluoro-5-trifluoromethyl pyridine-2-amine, 3-fluoro-5-trifluoromethyl-2-pyridinamine
PubChem CID 25067349
IUPAC Name 3-fluoro-5-(trifluoromethyl)pyridin-2-amine
SMILES C1=C(C=NC(=C1F)N)C(F)(F)F

Specifications

Melting Point 64°C to 68°C
Quantity 5 g
UN Number UN2811
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 2-Amino-3-fluoro-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.