CAS RN 852062-17-0
CAS RN 852062-17-0
IUPAC Name:
3-fluoro-5-(trifluoromethyl)pyridin-2-amine
Synonyms:
3-fluoro-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol):
180.11
Molecular Formula:
C6H4F4N2
InChi Key:
HAVVQRHBCUBQGG-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=C(C=N1)C(F)(F)F
1
–
2
of
2
results
1
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%
CAS: 852062-17-0 Molecular Formula: C6H4F4N2 Molecular Weight (g/mol): 180.106 MDL Number: MFCD10699117 InChI Key: HAVVQRHBCUBQGG-UHFFFAOYSA-N Synonym: 2-amino-3-fluoro-5-trifluoromethyl pyridine,3-fluoro-5-trifluoromethyl pyridin-2-amine,2-amino-3-fluoro-5-trifluoromethylpyridine,3-fluoro-5-trifluoromethyl-2-pyridylamine,3-fluoro-5-trifluoromethyl pyridine-2-amine,3-fluoro-5-trifluoromethyl-2-pyridinamine PubChem CID: 25067349 IUPAC Name: 3-fluoro-5-(trifluoromethyl)pyridin-2-amine SMILES: C1=C(C=NC(=C1F)N)C(F)(F)F