missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-diiodopyridine, 95%, Thermo Scientific™

Product Code. 15423707
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423707 250 mg 250mg
11421770 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423707 Supplier Thermo Scientific Alfa Aesar Supplier No. H27511.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23597-15-1
Molecular Formula C5H4I2N2
Molecular Weight (g/mol) 345.91
MDL Number MFCD02663852
InChI Key QLJWHQFRDOOATP-UHFFFAOYSA-N
PubChem CID 350960
IUPAC Name 3,5-diiodopyridin-2-amine
SMILES C1=C(C=NC(=C1I)N)I

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Synonym 2-amino-3,5-diiodopyridine, 2-pyridinamine, 3,5-diiodo, acmc-1cip1, 2-pyridinamine,3,5-diiodo, 2-amino-3,5-diiodopyridine 250mg
Formula Weight 345.91
Percent Purity 95%
Chemical Name or Material 2-Amino-3,5-diiodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.