accessibility menu, dialog, popup

UserName

CAS RN 23597-15-1

NH2NII

CAS RN 23597-15-1

IUPAC Name: 3,5-diiodopyridin-2-amine
Synonyms: 3,5-diiodopyridin-2-amine
Molecular Weight (g/mol): 345.91
Molecular Formula: C5H4I2N2
InChi Key: QLJWHQFRDOOATP-UHFFFAOYSA-N
SMILES: NC1=NC=C(I)C=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results