missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Amino-2'-deoxyadenosine, 98%

Product Code. 15456959
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456959 Supplier Thermo Scientific Chemicals Supplier No. J65532.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10414-81-0 | C10H14N6O3 | 266.26 g/mol

2'-AdA can be used as starting material for the synthesis of 2'-amino nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-AdA can be used as starting material for the synthesis of 2′-amino nucleotides.

Solubility
Soluble in DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10414-81-0
Molecular Formula C10H14N6O3
Molecular Weight (g/mol) 266.26
MDL Number MFCD06657636
InChI Key CQKMBZHLOYVGHW-GPGUAWMRNA-N
Synonym 2'-amino-2'-deoxyadenosine, 2-amino-adenosine, 2'-deoxy-2'-aminoadenosine, 2r,3s,4r,5r-4-amino-5-6-aminopurin-9-yl-2-hydroxymethyl oxolan-3-ol, 4-amino-5-6-amino-purin-9-yl-2-hydroxymethyl-tetrahydro-furan-3-ol, 2ad, 2-amino-2-deoxyadenosine, adenosine, 2'-amino,-2'-deoxy, 2-nh2-da, adenosine,2'-amino-2'-deoxy
PubChem CID 447594
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in DMSO.
IUPAC Name (2R,3S,4R,5R)-4-amino-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 266.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Amino-2'-deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.