missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate, 97%, Thermo Scientific™

Product Code. 10713803
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713803 1 g Amber Glass Bottle 1g
10151202 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713803 Supplier Thermo Scientific Maybridge Supplier No. BTB13907DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5425-81-0
Molecular Formula C8H9ClN2O3
Molecular Weight (g/mol) 216.621
MDL Number MFCD00025591
InChI Key UJBOVJRECBNSDI-UHFFFAOYSA-N
Synonym 2-amino-1-4-nitrophenyl ethanone hydrochloride, 2-amino-4'-nitro acetophenone hydrochloride, 2-amino-p-nitro-acetophenone hcl, 2-amino-p-nitro-acetophenone hydrochloride, 2-amino-1-4-nitro-phenyl ethanone hydrochloride, 2-amino-1-4-nitro-phenyl-ethanone hydrochloride, ethanone, 2-amino-1-4-nitrophenyl-, monohydrochloride, 2-amino-1-4-nitrophenyl ethan-1-one, chloride, 4-nitrophenacylamine hydrochloride hydrate, aronis24405
PubChem CID 2792801
IUPAC Name 2-amino-1-(4-nitrophenyl)ethanone;hydrochloride
SMILES C1=CC(=CC=C1C(=O)CN)[N+](=O)[O-].Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula HCl·H2O
Quantity 1 g
Formula Weight 234.64
Percent Purity 97%
Chemical Name or Material 2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.