accessibility menu, dialog, popup

UserName

ClNH2ONOO

CAS RN 5425-81-0

IUPAC Name: 2-amino-1-(4-nitrophenyl)ethan-1-one hydrochloridyl
Synonyms: 2-amino-1-(4-nitrophenyl)ethanone hydrochloridyl
Molecular Weight (g/mol): 215.61
Molecular Formula: C8H8ClN2O3
InChi Key: PFFDYNXIXWUUIM-UHFFFAOYSA-N
SMILES: [Cl].NCC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)