missing translation for 'onlineSavingsMsg'
Learn More

2-(Acetylamino)-N-(phenylmethyl)propanamide, TRC

Product Code. 30006640
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006640 500 mg 500mg
30006362 50 mg 50mg
30005451 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006640 Supplier Toronto Research Chemicals Supplier No. TRCA173725500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108258-31-7
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.14
Synonym 2-(Acetylamino)-N-(phenylmethyl)propanamide, 2-(Acetylamino)-N-(phenylmethyl)propanamide;
IUPAC Name methyl D-asparaginate
SMILES NC(C[C@@H](N)C(OC)=O)=O

Specifications

Analyte or Component Names 2-(Acetylamino)-N-(phenylmethyl)propanamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H10N2O3/c1-10-5(9)3(6)2-4(7)8/h3H,2,6H2,1H3,(H2,7,8)/t3-/m1/s1
Solution Type Neat
Industry Type Forensics
Quantity 500 mg
Formula Weight 146.07
Chemical Name or Material 2-(Acetylamino)-N-(phenylmethyl)propanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.