CAS RN 108258-31-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-(Acetylamino)-N-(phenylmethyl)propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Acetylamino)-N-(phenylmethyl)propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Acetylamino)-N-(phenylmethyl)propanamide, TRC
CAS: 108258-31-7 Molecular Formula: C5H10N2O3 Molecular Weight (g/mol): 146.14 Synonym: 2-(Acetylamino)-N-(phenylmethyl)propanamide,2-(Acetylamino)-N-(phenylmethyl)propanamide; IUPAC Name: methyl D-asparaginate SMILES: NC(C[C@@H](N)C(OC)=O)=O