missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific™

Product Code. 11387865
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11387865

Brand: Thermo Scientific Alfa Aesar L02849.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55620-18-3
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00014375
InChI Key UXOWQQCLBQBRMQ-VOTSOKGWSA-N
Synonym tylmarin, o-acetoxycinnamic acid, 2-acetoxycinnamic acid, o-acetyl-o-coumaric acid, cinnamic acid, 2-acetoxy, cinnamic acid, o-hydroxy-, acetate, 3-2-acetyloxy phenyl-2-propenoic acid, e-3-2-acetyloxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2-acetyloxy phenyl
PubChem CID 5372859
IUPAC Name (E)-3-(2-acetyloxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 154°C to 158°C
Quantity 1 g
Beilstein 2694007
Formula Weight 206.2
Percent Purity ≥98%
Chemical Name or Material 2-Acetoxycinnamic acid, predominantly trans
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.