missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals

Product Code. 11470207
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470207 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11470207 Supplier Thermo Scientific Alfa Aesar Supplier No. B21646.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-16-8
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00049100
InChI Key KEVMYFLMMDUPJE-UHFFFAOYSA-N
Synonym octane, 2,7-dimethyl, diisoamyl, 2,7dimethyloctane, diisopentyl, octane,7-dimethyl, 2,7-dimethyl-octane, acmc-1bq62
PubChem CID 14070
IUPAC Name 2,7-dimethyloctane
SMILES CC(C)CCCCC(C)C

Specifications

Melting Point -55°C
Density 0.72
Boiling Point 159°C to 160°C
Linear Formula (CH3)2CH(CH2)4CH(CH3)2
Refractive Index 1.409
Quantity 1 g
UN Number UN3295
Formula Weight 142.29
Percent Purity 98%
Chemical Name or Material 2,7-Dimethyloctane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.