Learn More
2,6-Pyridinedicarboxylic Acid, 99%, Thermo Scientific Chemicals
£6.75 - £158.00
Chemical Identifiers
CAS | 499-83-2 |
---|---|
Molecular Weight (g/mol) | 167.12 |
MDL Number | MFCD00006299 |
InChI Key | WJJMNDUMQPNECX-UHFFFAOYSA-N |
Synonym | 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid |
PubChem CID | 10367 |
ChEBI | CHEBI:46837 |
IUPAC Name | pyridine-2,6-dicarboxylic acid |
SMILES | C1=CC(=NC(=C1)C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10705791
|
Thermo Scientific Acros
131880050 |
5 g | |||||||
10573681
|
Thermo Scientific Acros
131880250 |
25 g |
£20.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10103570
|
Thermo Scientific Acros
131881000 |
100 g |
£49.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10064043
|
Thermo Scientific Acros
131885000 |
500 g |
£158.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
499-83-2 | |
MFCD00006299 | |
2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid | |
CHEBI:46837 | |
C1=CC(=NC(=C1)C(=O)O)C(=O)O |
167.12 | |
WJJMNDUMQPNECX-UHFFFAOYSA-N | |
10367 | |
pyridine-2,6-dicarboxylic acid |
Specifications
499-83-2 | |
100.0 | |
Authentic | |
C7H5NO4 | |
MFCD00006299 | |
2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid | |
C1=CC(=NC(=C1)C(=O)O)C(=O)O | |
167.12 | |
CHEBI:46837 | |
98.5 to 101.5% | |
2, 6-Pyridinedicarboxylic acid |
98.5 | |
White | |
99% | |
(C5H3N)(COOH)2 | |
22, 154 | |
WJJMNDUMQPNECX-UHFFFAOYSA-N | |
pyridine-2,6-dicarboxylic acid | |
10367 | |
167.12 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only