missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxylic Acid, 99%

Product Code. 10705791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10705791 5 g 5g
10573681 25 g 25g
10103570 100 g 100g
10064043 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10705791 Supplier Thermo Scientific Acros Supplier No. 131880050
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 499-83-2 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-83-2
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006299
InChI Key WJJMNDUMQPNECX-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
PubChem CID 10367
ChEBI CHEBI:46837
IUPAC Name pyridine-2,6-dicarboxylic acid
SMILES C1=CC(=NC(=C1)C(=O)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H5NO4
Linear Formula (C5H3N)(COOH)2
Quantity 5 g
Beilstein 22, 154
Formula Weight 167.12
Percent Purity 98.5 to 101.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Pyridinedicarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.