missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylpyridine, 97%

Product Code. 11420638
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420638 25 g 25g
11410638 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420638 Supplier Thermo Scientific Alfa Aesar Supplier No. B25179.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3558-69-8 | C17H13N | 231.298 g/mol

2,6-Diphenylpyridine is used as a tridentate [C?N?C] dianionic ligand in organogold(III) chemistry, an intermediate. It readily adds an electron from alkali metals in THF to give a radical anion which is a powerful reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diphenylpyridine is used as a tridentate [C∧N∧C] dianionic ligand in organogold(III) chemistry, an intermediate. It readily adds an electron from alkali metals in THF to give a radical anion which is a powerful reducing agent.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3558-69-8
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD00006284
InChI Key PJUOHDQXFNPPRF-UHFFFAOYSA-N
Synonym pyridine, 2,6-diphenyl, unii-zi75v15y1t, 2,6-diphenyl-pyridine, 2,6-diphenyl pyridine, maybridge3_000511, acmc-1cq41, pjuohdqxfnpprf-uhfffaoysa
PubChem CID 72920
IUPAC Name 2,6-diphenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 74°C to 81°C
Boiling Point 210°C (3 mmHg)
Quantity 25 g
Beilstein 7516
Solubility Information Insoluble in water.
Formula Weight 231.3
Percent Purity 97%
Chemical Name or Material 2,6-Diphenylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.