CAS RN 3558-69-8
IUPAC Name:
2,6-diphenylpyridine
Synonyms:
2,6-diphenylpyridine
Molecular Weight (g/mol):
231.3
Molecular Formula:
C17H13N
InChi Key:
PJUOHDQXFNPPRF-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=CC=CC(=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
2,6-Diphenylpyridine, 97%
CAS: 3558-69-8 Molecular Formula: C17H13N Molecular Weight (g/mol): 231.298 MDL Number: MFCD00006284 InChI Key: PJUOHDQXFNPPRF-UHFFFAOYSA-N Synonym: pyridine, 2,6-diphenyl,unii-zi75v15y1t,2,6-diphenyl-pyridine,2,6-diphenyl pyridine,maybridge3_000511,acmc-1cq41,pjuohdqxfnpprf-uhfffaoysa PubChem CID: 72920 IUPAC Name: 2,6-diphenylpyridine SMILES: C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3